Skip to Content
MilliporeSigma

A8751

Adenine hydrochloride hydrate

≥99%, powder

Synonym(s):

Adenine hydrochloride, Adenine,monohydrochloride, 6-Aminopurine hydrochloride hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H5N5 · HCl · xH2O
CAS Number:
Molecular Weight:
171.59 (anhydrous basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
MDL number:
Assay:
≥99%
Biological source:
synthetic
Form:
powder
Solubility:
warm water: 50 mg/mL, clear to hazy, colorless to faint yellow or tan
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Adenine hydrochloride hydrate, ≥99%

biological source

synthetic

Quality Segment

assay

≥99%

form

powder

solubility

warm water: 50 mg/mL, clear to hazy, colorless to faint yellow or tan

SMILES string

O.Cl.Nc1ncnc2[nH]cnc12

InChI

1S/C5H5N5.ClH.H2O/c6-4-3-5(9-1-7-3)10-2-8-4;;/h1-2H,(H3,6,7,8,9,10);1H;1H2

InChI key

MYRDTAUFFBYTHA-UHFFFAOYSA-N

General description

Adenine is a purine nucleobase with a wide range of chemical and biochemical roles in vivo and in vitro. It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD), and signaling molecules (cAMP).

Application

Adenine hydrochloride hydrate has been used:

  • as a supplement in a rich medium for galactose induction during yeast cell culture.
  • for the incubation of MiaPaCa2 cell lines for cell viability and co-culture assay.
  • as a component of the Pombe Glutamate (PMG)media for the germination of Schizosaccharomyces pombe.


Still not finding the right product?

Explore all of our products under Adenine hydrochloride hydrate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library