Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A8736

Sigma-Aldrich

A12B4C3

≥98% (HPLC)

Synonym(s):

2-(1-Hydroxyundecyl)-1-(4-nitrophenylamino)-6-phenyl-6,7a-dihydro-1H-pyrrolo[3,4-b]pyridine-5,7(2H,4aH)-dione, 4a,7a-Dihydro-2-(1-hydroxyundecyl)-1-[(4-nitrophenyl)amino]-6-phenyl-1H-pyrrolo[3,4-b]pyridine-5,7(2H,6H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H38N4O5
CAS Number:
Molecular Weight:
534.65
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

CCCCCCCCCCC(O)C1C=CC2C(N1Nc3ccc(cc3)[N+]([O-])=O)C(=O)N(C2=O)c4ccccc4

InChI

1S/C30H38N4O5/c1-2-3-4-5-6-7-8-12-15-27(35)26-21-20-25-28(30(37)32(29(25)36)23-13-10-9-11-14-23)33(26)31-22-16-18-24(19-17-22)34(38)39/h9-11,13-14,16-21,25-28,31,35H,2-8,12,15H2,1H3

InChI key

KVHGJAKTBPFFNV-UHFFFAOYSA-N

Application

A12B4C3 has been used as an inhibitor of polynucleotide kinase/phosphatase to study its effects on prostate cancer (PC-3) cells.

Biochem/physiol Actions

2-(1-hydroxyundecyl)-1- (4-nitrophenylamino)-6-phenyl-6,7a-dihydro-1Hpyrrolo [3,4-b]pyridine-5,7(2H,4aH)-dione (A12B4C3) is a polysubstituted piperidine, that is capable of altering the secondary structure of polynucleotide kinase 3′-phosphatase (PNKP). It functions as an allosterically regulated noncompetitive inhibitor of human PNKP phosphatase activity.
A12B4C3 is a potent and specific hPNKP phosphatase inhibitor. A12B4C3 doubles the radiosensitivity of human A549 lung carcinoma and MDA-MB-231 breast adenocarcinoma cells.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Yang et al.
Cell death and differentiation, 27(8), 2433-2450 (2020-03-08)
Cancer cells are defective in DNA repair, so they experience increased DNA strand breaks, genome instability, gene mutagenesis, and tumorigenicity; however, multiple classic DNA repair genes and pathways are strongly activated in malignant tumor cells to compensate for the DNA
A12B4C3, an inhibitor of polynucleotide kinase/phosphatase enhances radio-sensitivity in PC-3 cells exposed to carbon ion beam
Srivastava P, et al.
Journal of Biomechanical Science and Engineering null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service