Skip to Content
MilliporeSigma

A7973

[Arg13] Amyloid-β-Protein Fragment 1-40

≥80% (HPLC), lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Form:
lyophilized powder
Assay:
≥80% (HPLC)
Mol wt:
4350
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥80% (HPLC)

form

lyophilized powder

mol wt

4350

composition

Peptide Content, ≥70%

solubility

H2O: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)

Application

Used to study the effect of Arg13 on Aβ aggregation.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Suzuki et al.
The Journal of experimental medicine, 191(11), 1905-1920 (2000-06-06)
Shigella, the causative agent of bacillary dysentery, is capable of directing its movement within host cells by exploiting actin dynamics. The VirG protein expressed at one pole of the bacterium can recruit neural Wiskott-Aldrich syndrome protein (N-WASP), a downstream effector
L K Simmons et al.
Molecular pharmacology, 45(3), 373-379 (1994-03-01)
Amyloid beta peptide (A beta), the major protein constituent of senile plaques in patients with Alzheimer's disease, is believed to facilitate the progressive neurodegeneration that occurs in the latter stages of this disease. Early attempts to characterize the structure-activity relationship

Global Trade Item Number

SKUGTIN
A7973-.5MG04061833217429

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service