Skip to Content
MilliporeSigma

A7095

Amyloglucosidase from Aspergillus niger

≥260 U/mL, aqueous solution

Synonym(s):

1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
Specific activity:
≥260 U/mL
Biological source:
Aspergillus niger
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Aspergillus niger

Quality Segment

form

aqueous solution

specific activity

≥260 U/mL

density

~1.2 g/mL at 25 °C

storage temp.

2-8°C

General description

Stabilized with glucose.
Amyloglucosidase is a disaccharidase−type alpha-glucosidase, produced by several species of Aspergillus genus. Immobilization of amyloglucosidase is known to increased its stability.

Application

Amyloglucosidase from Aspergillus niger has been used in in vitro digestions. It has also been used in the isolation of insoluble and soluble dietary fibre from quinoa and amaranth.

Biochem/physiol Actions

Amyloglucosidase from Aspergillus niger is capable of hydrolyzing the α-D-(1-4), the α-D-(1-6), and the α-D-(1-3) glucosidic bonds of oligosaccharides. Amyloglucosidase is an extracellular enzyme that converts starch to dextrins and glucose. The enzyme is used in the starch-processing industry for the commercial production of D-glucose from corn syrups.

Legal Information

A product of Novozymes Corp.


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
A7095-50ML04061838251602