Skip to Content
MilliporeSigma

A6853

α2A-Adrenergic Receptor preparation

membrane suspension

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.84
MDL number:
Form:
membrane suspension
Biological source:
human
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

form

membrane suspension

shipped in

dry ice

storage temp.

−70°C

Biochem/physiol Actions

Membranes suspended in 50 mM Tris-HCl containing 10% sucrose, pH 7.65. Actual concentration and specific binding capacity are provided with each lot.


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
A6853-100UN04061831168495