Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352204
NACRES:
NA.83
Product Name
Alkaline Phosphatase Stop Solution, for ELISA
Quality Segment
form
powder
solubility
water: 100 mL, clear, colorless
storage temp.
2-8°C
Application
Stop solution for Alkaline Phosphatase Red and Blue microwell substrate solutions. Use 50 μl of stop solution per 200 μl of reaction mix. Alkaline Phosphatase Stop Solution has been used in terminating the alkaline phosphatase and nitrotetrazolium blue chloride (NBT)/5-bromo-4-chloro-3-indolyl phosphate (BCIP) reaction on nitrocellulose membrane.
Preparation Note
Reconstitute with 100 mL of water and mix for 30-60 min.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A5852-100ML | 04061838168429 |
