Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A5177

Aspirator tube assemblies for calibrated microcapillary pipettes

pkg of 5 ea

Synonym(s):

Aspirator Tube Assemblies, Microcapillary Pipettes, Tube Assemblies for Pipettes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

packaging

pkg of 5 ea

Related Categories

General description

Consists of flexible silicone rubber nosepiece, hard plastic mouthpiece, and 15 inches of latex tubing.

Application

Aspirator tube assemblies for calibrated microcapillary pipettes have been used:
  • as a mouthpiece for pipettes to gently suction out plasmid solution
  • to aid during intrathoracic injection using a glass needle
  • as a mouth pipette attached to glass needles for transferring melanocytes to the culture dishes


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juntang Lin et al.
The Journal of comparative neurology, 524(5), 940-962 (2015-08-20)
Pax3 and Pax7 are closely related transcription factors that are widely expressed in the developing nervous system and somites. In the CNS, both genes are expressed in the dorsal part of the neural tube during development. Pax3 and Pax7 are
Juntang Lin et al.
The Journal of comparative neurology, 525(7), 1618-1632 (2016-11-20)
Pax3 is a transcription factor that belongs to the paired box family. In the developing spinal cord it is expressed in the dorsal commissural neurons, which project ascending axons contralaterally to form proper spinal cord-brain circuitry. While it has been
Taeyong Kim et al.
Scientific reports, 8(1), 12060-12060 (2018-08-15)
Regenerative therapeutic approaches for myocardial diseases often involve delivery of stem cells expanded ex vivo. Prior studies indicate that cell culture conditions affect functional and phenotypic characteristics, but relationship(s) of cultured cells derived from freshly isolated populations and the heterogeneity
Suhanya Duraiswamy et al.
Infection and immunity, 86(4) (2018-01-31)
Urinary tract infections (UTIs) are a major infection of humans, particularly affecting women. Recurrent UTIs can cause significant discomfort and expose patients to high levels of antibiotic use, which in turn contributes to the development of higher antibiotic resistance rates.
Bo Feng et al.
BMC neuroscience, 6, 46-46 (2005-07-16)
Rag1 (Recombination activation gene-1) mediates genomic rearrangement and is essential for adaptive immunity in vertebrates. This gene is also expressed in the olfactory epithelium, but its function there is unknown. Using a transgenic zebrafish line and immunofluorescence, we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service