Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4929

Supelco

N,N′-Bis(acryloyl)cystamine

BioReagent, suitable for electrophoresis

Synonym(s):

2,2′-(Bisacrylamino)diethyl disulfide, BAC, Bis(2-acrylamidoethyl) disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O2S2
CAS Number:
Molecular Weight:
260.38
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

form

powder

suitability

suitable for electrophoresis

storage temp.

−20°C

SMILES string

C=CC(=O)NCCSSCCNC(=O)C=C

InChI

1S/C10H16N2O2S2/c1-3-9(13)11-5-7-15-16-8-6-12-10(14)4-2/h3-4H,1-2,5-8H2,(H,11,13)(H,12,14)

InChI key

DJVKJGIZQFBFGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min.
BAC is used as a reversible cross-linker for polyacrylamide gels. The disulfide linkage can be broken with a suitable reducing agent. A 12:1 ratio of acrylamide:BAC is recommended for DNA purification. A one gram slice of a 4% gel will dissolve in 5 mL of 100 mM DTT, TBE solution (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nick Orr et al.
Human molecular genetics, 24(10), 2966-2984 (2015-02-06)
We recently identified a novel susceptibility variant, rs865686, for estrogen-receptor positive breast cancer at 9q31.2. Here, we report a fine-mapping analysis of the 9q31.2 susceptibility locus using 43 160 cases and 42 600 controls of European ancestry ascertained from 52
Hiroko Sano et al.
PLoS genetics, 11(5), e1005209-e1005209 (2015-05-29)
The coordination of growth with nutritional status is essential for proper development and physiology. Nutritional information is mostly perceived by peripheral organs before being relayed to the brain, which modulates physiological responses. Hormonal signaling ensures this organ-to-organ communication, and the
Na-Yu Chia et al.
Gut, 64(5), 707-719 (2014-07-24)
Gastric cancer (GC) is a deadly malignancy for which new therapeutic strategies are needed. Three transcription factors, KLF5, GATA4 and GATA6, have been previously reported to exhibit genomic amplification in GC. We sought to validate these findings, investigate how these
Kevin R Bushell et al.
Blood, 125(6), 999-1005 (2014-12-04)
Non-Hodgkin lymphomas (NHLs) are the most common cancer to affect pet dogs. In contrast to the many genes whose mutation contributes to lymphomagenesis in humans, relatively little is known about the acquired genetic alterations that lead to canine B-cell lymphomas
Sehrish Rafique et al.
Genome biology, 16, 145-145 (2015-08-04)
Epigenetic changes are being increasingly recognized as a prominent feature of cancer. This occurs not only at individual genes, but also over larger chromosomal domains. To investigate this, we set out to identify large chromosomal domains of epigenetic dysregulation in

Articles

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service