Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
Form:
lyophilized powder
Assay:
≥90% (HPLC)
Mol wt:
1324
Application
Aβ fragment used to study aggregation.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
H Inouye et al.
Biophysical journal, 64(2), 502-519 (1993-02-01)
To elucidate the relation between amyloid fibril formation in Alzheimer disease and the primary structure of the beta/A4 protein, which is the major component of the amyloid, we have been investigating the ability of peptides sharing sequences with beta/A4 to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service