Skip to Content
MilliporeSigma

A4566

L-Arginine p-nitroanilide dihydrochloride

protease substrate, ≥99.0% (TLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H18N6O3 · 2HCl
CAS Number:
Molecular Weight:
367.23
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Arginine p-nitroanilide dihydrochloride, protease substrate

Quality Segment

assay

≥99.0% (TLC)

form

powder

solubility

DMSO:methanol (1:1): 49.00-51.00 mg/mL, clear, pale yellow to yellow

storage temp.

2-8°C

SMILES string

Cl.N[C@@H](CCCNC(N)=N)C(=O)Nc1ccc(cc1)N(=O)=O

InChI

1S/C12H18N6O3.ClH/c13-10(2-1-7-16-12(14)15)11(19)17-8-3-5-9(6-4-8)18(20)21;/h3-6,10H,1-2,7,13H2,(H,17,19)(H4,14,15,16);1H/t10-;/m0./s1

InChI key

KBODDOUNTSALJF-PPHPATTJSA-N

Biochem/physiol Actions

p-Nitroanilides derivatives of amino acids are mostly used as chromogenic substrates for measuring the activity of proteolytic enzymes.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
A4566-100MG04061833374702
A4566-250MG04061833374719