Skip to Content
MilliporeSigma

A4394

N-Acetyl-D-glucosamine 6-phosphate sodium salt

≥98% (TLC)

Synonym(s):

GlcNAc-6-P Sodium Salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H16NO9P · xNa+
Molecular Weight:
301.19 (free acid basis)
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

natural (inorganic)

Quality Segment

assay

≥98% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

impurities

<10% water (Karl Fischer)

color

white to light yellow

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na].CC(=O)NC1C(O)OC(COP(O)(O)=O)C(O)C1O

InChI

1S/C8H16NO9P.Na.H/c1-3(10)9-5-7(12)6(11)4(18-8(5)13)2-17-19(14,15)16;;/h4-8,11-13H,2H2,1H3,(H,9,10)(H2,14,15,16);;

InChI key

AILBPHCGSZBCSM-UHFFFAOYSA-N

Application

N-Acetyl-D-glucosamine 6-phosphate (GlcNAc-6P), an intracellular form of GlcNAc, is a substrate used to identify, differentiate and characterize N-acetyl-D-glucosamine-phosphate deacetylase(s).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library