Skip to Content
MilliporeSigma

A3832

Adrenomedullin Fragment 22-52 human

≥97% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C159H252N46O48
CAS Number:
Molecular Weight:
3575.98
UNSPSC Code:
12352209
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Adrenomedullin Fragment 22-52 human, ≥97% (HPLC)

Quality Segment

assay

≥97% (HPLC)

form

powder

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ADM(133)

Biochem/physiol Actions

Adrenomedullin Fragment 22-52 [ADM22-52] is used as an adrenomedullin (ADM) receptor antagonist to study the functions and mechanism of action of ADM signaling. ADM(22-52) is used to differentiate adrenomedullin binding sites in various cells and tissues.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library