Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

A3281

Sigma-Aldrich

N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride

Synonym(s):

W-7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21ClN2O2S · HCl
CAS Number:
Molecular Weight:
377.33
Beilstein:
6030174
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Mode of action

cell membrane | interferes
enzyme | inhibits

storage temp.

−20°C

SMILES string

Cl[H].NCCCCCCNS(=O)(=O)c1cccc2c(Cl)cccc12

InChI

1S/C16H21ClN2O2S.ClH/c17-15-9-5-8-14-13(15)7-6-10-16(14)22(20,21)19-12-4-2-1-3-11-18;/h5-10,19H,1-4,11-12,18H2;1H

InChI key

OMMOSRLIFSCDBL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride has been used to study its effect of on coated pit staining.

Biochem/physiol Actions

N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide (W-7) is a calmodulin antagonist. It can prevent calcium-dependent protein kinase, that is sensitive to phospholipid.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Osawa et al.
FEBS letters, 442(2-3), 173-177 (1999-02-03)
Small-angle X-ray scattering and nuclear magnetic resonance were used to investigate the structural change of calcium-bound calmodulin (Ca2+/CaM) in solution upon binding to its antagonist, N-(6-aminohexyl)-5-chloro-1-naphthalenesulfonamide (W-7). The radius of gyration was 17.4+/-0.3 A for Ca2+/CaM-W-7 with a molar ratio
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide (W-7), a calmodulin antagonist, also inhibits phospholipid sensitive calcium-dependent protein kinase
Schatzman RC, et al.
Biochimica et biophysica acta. General subjects, 755(1), 144-147 (1983)
Matteo Coen et al.
The American journal of pathology, 183(3), 996-1009 (2013-07-11)
Several observations suggest the expansion of a distinct medial smooth muscle cell (SMC) subset in atherosclerosis and restenosis. We characterized the phenotypic features of SMC subsets in cultures derived from human carotid endarterectomy specimens. Specimens comprised an undiseased portion (thin
Valentin Jaumouillé et al.
Channels (Austin, Tex.), 5(4), 308-313 (2011-06-10)
NHE3 is regulated via alterations in membrane surface charge. This is achieved through altered binding of cationic regions in the cytosolic-terminus of the exchanger with the inner leaflet of the plasma membrane. Calmodulin antagonists, including W-7, regulate surface potential and
L H Wang et al.
The Journal of cell biology, 123(5), 1107-1117 (1993-12-01)
The clathrin-coated pit lattice is held onto the plasma membrane by an integral membrane protein that binds the clathrin AP-2 subunit with high affinity. In vitro studies have suggested that this protein controls the assembly of the pit because membrane

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service