Skip to Content
MilliporeSigma

A301000

DMT-dA(bz)-CPG

pore size 500 Å, 30-40μmol/g

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
12352200
Assay:
≥98.0% (Nucleoside Purity, reversed phase HPLC)
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

type

for DNA synthesis

product line

Proligo Reagents

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

pore size

500 Å

capacity

≥99 % (Coupling Efficiency)

λ

conforms (UV/VIS Identity)

storage temp.

2-8°C



Still not finding the right product?

Explore all of our products under DMT-dA(bz)-CPG


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library