Skip to Content
MilliporeSigma

A2598

ART® self-sealing barrier pipette tips

ART 10, volume range 0.1-10 μL, sterile

Synonym(s):

pipet tips, pipette tips, MBP 2139, dispensing tip, liquid transfer tip

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.01
UNSPSC Code:
41121601
Volume:
0.1—10 μL
Sterility:
sterile
Feature:
DNase free, RNase free, barrier
Packaging:
case of 5 packs, pack of 960 ea (racked)
Manufacturer/tradename:
Molecular BioProducts 2139
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

colorless

sterility

sterile

feature

DNase free, RNase free, barrier

packaging

case of 5 packs, pack of 960 ea (racked)

manufacturer/tradename

Molecular BioProducts 2139

technique(s)

PCR: suitable

maximum volume

10 μL

volume range

0.1-10 μL

volume

0.1—10 μL

compatibility

for use with

  • Biohit mLINE .5-10 μl
  • Biohit Electronic .2-10 μl
  • Biohit Variable Single/Multi-Channel .5-10 μl
  • Capp Aero 10 μl
  • Corning 4960 .5-10 μl
  • Finnpipette 4500 Digital .2-2 μl
  • Finnpipette 4500 Digital Micro .5-10 μl
  • Finnpipette Focus 4600 3 μl, 5 μl, 10 μl
  • Finnpipette Multi-Channel .5-10 μl
  • Hamilton SoftGrip Precision 2 μl/10 μl
  • Nichiryo BenchMate .2-2 μl/.5-10 μl
  • Pipetman P-2 .1-2 μl
  • Pipetman P-10 .5-10 μl
  • Rainin EDP 2 .5-10 μl
  • Rainin EDP Plus .5-10 μl
  • Rainin Pipet Plus .2-2 μl/.5-10 μl
  • Rainin Pipetman .2-2 μl/.5-10 μl
  • Transferpette Single/Multi-Channel .5-10 μl/2-20 μl
  • Transferpette Ultra-Micro 2-20 μl

General description

2 μL reference mark; the perfect tip for pipetting low volumes

Application

ART® self-sealing barrier pipette tips have been used for the quantitative profiling of endocannabinoids in lipoproteins by LC–MS (liquid chromatography–mass spectrometry)/MS. They have also been used for RNA isolation and reverse transcription.

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library