Skip to Content
MilliporeSigma

A2350

AlumaSeal® II film

non-sterile

Synonym(s):

Sealing film

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41122410
Manufacturer/tradename:
Excel Scientific AF-100
Material:
aluminum film
W × l:
8.26 cm × 14.61 cm
Sterility:
non-sterile
Packaging:
pkg of one zippered bag of 100
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

aluminum film

sterility

non-sterile

feature

piercable

manufacturer/tradename

Excel Scientific AF-100

packaging

pkg of one zippered bag of 100

parameter

-80 to +120 °C temp. range

technique(s)

PCR: suitable

L

146.1 mm

W × L

8.26 cm × 14.61 cm

W

82.6 mm

foreign activity

RNase/ DNase, certified free

General description

A soft aluminum foil film designed for the sealing of PCR plates minimizing evaporation and vapor loss during thermal cycling. The film pierces very easily for direct sample recovery with a multichannel pipettor or robotic metal probes without risk of aerosols, cross-contamination or significant gumming of adhesive. An extremely strong, medical-grade adhesive provides a uniform, complete seal which can withstand PCR thermal cycling. Recommended for temperatures from -80 °C to 120 °C, measures 82.6 mm x 146.1 mm and offers sufficient sealing area for all PCR plates. A 36 μm non-permeable soft aluminum foil film eliminates the need for heat-sealing devices or mats during thermal cycling. The proprietary adhesive can withstand harsh chemicals, solvents and reagents. Two end tabs for easy and accurate positioning and labeling.

Application

To seal multiplate used for:
  • PCR
  • Long-term storage
  • Light sensitive assays
  • Robotics

Features and Benefits

  • minimizes evaporation and vapor loss during thermal cycling
  • easily pierced for direct sample recovery without risk of aerosols or cross-contamination
  • strong adhesive provides a uniform, complete seal that withstands PCR thermal cycling
  • proprietary adhesive withstands harsh chemicals, solvents and reagents
  • two end tabs for easy and accurate positioning and labeling

Legal Information

AlumaSeal is a registered trademark of Excel Scientific, Inc.


Still not finding the right product?

Explore all of our products under AlumaSeal® II film




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library