Skip to Content
MilliporeSigma

A2221

Antithrombin III from human plasma

lyophilized powder, ≥95% (SDS-PAGE)

Synonym(s):

Factor Xa inhibitor, Heparin cofactor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
290-516-4
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human plasma

Quality Segment

assay

≥95% (SDS-PAGE)

form

lyophilized powder

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... SERPINC1(462)

Biochem/physiol Actions

Serine protease inhibitor; Inhibitor of thrombin and activated factor X (Xa). Effective concentration equimolar with proteinase.

Physical form

Lyophilized from 20 mM Tris, pH 8.0

Other Notes

Not to be confused with thrombin antiserum.
One unit will form a complex and inhibit one unit of bovine thrombin at 25 degree C, using a 5 minute incubation in the presence of heparin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library