Skip to Content
MilliporeSigma

A2025

DL-Aspartic acid potassium salt

≥98% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H6NO4K
CAS Number:
Molecular Weight:
171.19
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
238-407-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

DL-Aspartic acid potassium salt,

assay

≥98% (TLC)

form

powder

color

white

solubility

H2O: soluble

application(s)

cell analysis
peptide synthesis

SMILES string

[K+].NC(CC(O)=O)C([O-])=O

InChI

1S/C4H7NO4.K/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H,6,7)(H,8,9);/q;+1/p-1

InChI key

TXXVQZSTAVIHFD-UHFFFAOYSA-M

Biochem/physiol Actions

DL-Aspartic acid (DL-Asp) is a racemic mixture of the proteinogenic amino acid L-aspartate and the non-proteinogenic amino acid D-aspartate. DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. DL-Aspartic acid is used to develop and assess amino acid racemic resolution and racemic interconversion technologies. Principal neurotransmitter for fast synaptic excitation.
Principal neurotransmitter for fast synaptic excitation.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library