Skip to Content
MilliporeSigma

A1276

Amastatin hydrochloride hydrate

powder, ≥97% (HPLC)

Synonym(s):

(2S,3R)-3-Amino-2-hydroxy-5-methylhexanoyl-Val-Val-Asp hydrochloride hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H38N4O8 · HCl · xH2O
CAS Number:
Molecular Weight:
511.01 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Beilstein/REAXYS Number:
8888370
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Amastatin hydrochloride hydrate, ≥97% (HPLC)

biological source

synthetic (organic)

Quality Segment

assay

≥97% (HPLC)

form

powder

mp

202-205 °C

solubility

methanol: 1 mM (Stock solution stable for 1 month at −20 °C. Working solution stable for 1 day.)

antibiotic activity spectrum

neoplastics

mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

Cl[H].[H]O[H].CC(C)C[C@@H](N)[C@H](O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C21H38N4O8.ClH.H2O/c1-9(2)7-12(22)17(28)20(31)25-16(11(5)6)19(30)24-15(10(3)4)18(29)23-13(21(32)33)8-14(26)27;;/h9-13,15-17,28H,7-8,22H2,1-6H3,(H,23,29)(H,24,30)(H,25,31)(H,26,27)(H,32,33);1H;1H2/t12-,13+,15+,16+,17+;;/m1../s1

InChI key

WBYDQJQQTMJMQH-AFROIDPFSA-N

General description

Chemical structure: peptide

Biochem/physiol Actions

Amastatin is a slow, tight-binding inhibitor of aminopeptidases. It inhibits cytosolic leucine aminopeptidase (EC.3.4.11.1), microsomal aminopeptidase M (EC.3.4.11.2) and bacterial leucine aminopeptidase (EC.3.4.11.10). It is less effective against aminopeptidase A (EC 3.4.11.7), the enzyme that converts Angiotensin II to Angiotensin III. Effective concentration: 1-10 μM.
It inhibits cytosolic leucine aminopeptidase (EC.3.4.11.1), microsomal aminopeptidase M (EC.3.4.11.2) and bacterial leucine aminopeptidase (EC.3.4.11.10). It is less effective against aminopeptidase A (EC 3.4.11.7), the enzyme that converts angiotensin II to angiotensin III. Potentiates the CNS effects of vasopressin and oxytocin in vivo.

Effective concentration: 1-10 μM.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
A1276-25MG04061833257876
A1276-1MG04061833342022
A1276-5MG04061833257883
A1276-10MG04061833267080
A1276-.5MG04061833257869