Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1052

Sigma-Aldrich

Anti-Gliadin (Wheat)–Peroxidase Conjugate antibody produced in rabbit

fractionated antiserum, PBS solution

Synonym(s):

Anti-Gliadin (Wheat)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

PBS solution

species reactivity

oats, rye, soy, barley
wheat

should not react with

rice, potato

technique(s)

direct ELISA: 1:1,000
dot blot: 1:500
immunoblotting: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Gliadin is a class of glutamine containing, alcohol soluble proteins separable from wheat and rye glutens known as the prolamins.

Immunogen

native and heat-treated wheat gliadin

Application

Rabbit anti-gliadin (wheat)-peroxidase conjugate antibody can be used for immunoblot and ELISA(1:1000).

Biochem/physiol Actions

Gliadin is a class of glutamine containing, alcohol soluble proteins separable from wheat and rye glutens known as the prolamins. These proteins are associated with the harmful effects of celiac disease and gluten sensitive enteropathy in humans, causing characteristic changes in the small intestinal mucosa. In such cases, strict compliance to a gluten-free diet is usually recommended.
Gliadin proteins are associated with the harmful effects of celiac disease and gluten sensitive enteropathy in humans, causing characteristic changes in the small intestinal mucosa. In such cases, strict compliance to a gluten-free diet is usually recommended.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% MIT

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein characteristics that affect the quality of vital wheat gluten to be used in baking: A review
Ortolan F and Steel CJ
Comprehensive Reviews in Food Science and Food Safety, 16(3), 369-381 (2017)
Evaluation of the applicability of two different immunoassays for the detection of wheat gluten in baby foods.
Avila, C., & Quesada, A. R.
Biochemistry and Molecular Biology Education, 28(5), 261-264 (2000)
Celiac Disease: celiac sprue, gluten-sensitive enteropathy
Gheller-Rigoni AI, et al.
Clinical Medicint and Research, 2(1), 71-72 (2004)
W Weiss et al.
Electrophoresis, 14(8), 805-816 (1993-08-01)
In order to identify and characterize the wheat grain allergens involved in bakers' asthma, proteins were sequentially extracted from whole-meal flour. The polypeptide composition of the individual solubility fractions (albumin/globulin, gliadin and glutenin) was analyzed by sodium dodecyl sulfate-gel electrophoresis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service