Skip to Content
MilliporeSigma

A0919

Mouse IgG−Agarose

(Suspension in 0.5 M NaCl containing preservative.)

Synonym(s):

IgG-Agarose Beads

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
agarose conjugate
Technique(s):
immunoprecipitation (IP): suitable
Application:
IP
Citations:
92
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

agarose conjugate

Quality Segment

antibody form

purified immunoglobulin

form

(Suspension in 0.5 M NaCl containing preservative.)

extent of labeling

1 mg per mL

technique(s)

immunoprecipitation (IP): suitable

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix spacer

1 atom

storage temp.

2-8°C

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice.

Application

IgG-Agarose can be used as an immunoadsorbent. It can also be used to affinity purify antibodies, remove species specific cross-reacting antibodies or remove contaminating antibodies from an antiserum preparation. Typically cross-reacting antibodies may be removed from an antiserum preparation using an equal volume of IgGAgarose (resin volume). Mouse IgG-Agarose has been used for several immunoprecipitation assays,.
Mouse IgGs conjugated to agarose beads were used in immunoprecipitation assays to pre-clear HEK 293 whole cell extracts. Beads were incubated with extracts for 1 hour rotating at 4°C.

Physical form

Suspension in 0.5 M NaCl containing preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library