Skip to Content
MilliporeSigma

95284

Water

for molecular biology, suitable for FISH

Synonym(s):

H2O

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
NACRES:
NA.31
PubChem Substance ID:
UNSPSC Code:
12191602
EC Number:
231-791-2
MDL number:
Beilstein/REAXYS Number:
2050024
Grade:
Molecular Biology
Technique(s):
FISH: suitable
Bp:
100 °C (lit.)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Water, Molecular Biology, sterile filtered

grade

Molecular Biology

Quality Segment

sterility

sterile-filtered

form

liquid

quality

sterile filtered

shelf life

limited shelf life, expiry date on the label

technique(s)

FISH: suitable

impurities

DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected

refractive index

n20/D 1.34 (lit.)

pH

5-7

conductivity

0.0-5 μS/cm at 25 °C

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

cation traces

Ca: <10 ppb, Mg: <10 ppb

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

General description

Molecular biology grade water is sterile filtered and ready to use, free from DNase, RNase, phosphatase, and protease contamination.

Application

Water, sterile filtered has been used:

  • for cloning and plasmid construction.
  • for restriction digestion of PCR amplicons in polysaccharide A (PSA) restriction method.
  • in RNA isolation.
  • fluorescence in situ hybridization (FISH).
  • to dilute the samples of CVF (cervicovaginal fluid) and menstrual extracts for testing specificity through reverse transcriptase-polymerase chain reaction (RT-PCR) amplification of mRNA.
  • for DNA quantification using quantitative polymerase chain reaction (qPCR).


Still not finding the right product?

Explore all of our products under Water


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library