Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94280

Sigma-Aldrich

Urease from Canavalia ensiformis (Jack bean)

powder, ~1 U/mg

Synonym(s):

Jack bean urease, Urea amidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

form

powder

specific activity

~1 U/mg

mol wt

Mr ~480000

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Subunit molecular weight: ~90,770
Composed of six subunits with total molecular weight: ~544,620
Urease (Ure) enzyme is present in the cytosol. It is active at acidic pH and is expressed at 28°C. Urease consists of nickel and is present in archaea, bacteria, unicellular eukaryotes and plants.

Application

Application in (selective) hydrolysis/condensations;citation structurecitationcitation.
Urease from Canavalia ensiformis may be used for urea determination of various samples, such as legumes . It may be useful for the detection of pathogens as well as heavy-metal ions.
Urease from Canavalia ensiformis (Jack bean) has been used in purification and crystallization studies.

Biochem/physiol Actions

Urease catalyzes the hydrolysis of urea into carbon dioxide and ammonia. Urease is involved in nitrogen metabolism and urea degradation. Urease from Canavalia ensiformis binds 2 nickel ions per subunit .

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol urea (Cat. No. 51459) per minute at pH 8.0 and 25 °C.

Other Notes

Application in (selective) hydrolysis/condensations; structure.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I.A. Natchev
Tetrahedron, 44, 1511-1511 (1988)
Isothermal titration calorimetry to characterize enzymatic reactions
Methods in Enzymology, 567, 215-236 (2016)
M J Sculley et al.
Biophysical chemistry, 19(1), 39-47 (1984-01-01)
The use of bifunctional reagents to form cross-links between subunits in protein oligomers and subsequent disruption of noncovalent interactions with SDS allows comment upon the number of subunits and the symmetry in the original assembly. In existing treatments the number
Yersinia enterocolitica
Molecular Medical Microbiology, 1319-1344 (2015)
M.C.N. Jayasuriya et al.
Anim. Feed Sci. Technol., 8, 271-271 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service