Skip to Content
MilliporeSigma

93348

Trizma® phosphate monobasic

BioXtra, ≥99.0% (NT)

Synonym(s):

Mono(tris[hydroxymethyl]aminomethane) phosphate, Mono-Tris phosphate, Tris phosphate, Tris(hydroxymethyl)aminomethane phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2C(CH2OH)3 · H3PO4
CAS Number:
Molecular Weight:
219.13
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161700
EC Number:
230-260-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioXtra

Quality Segment

assay

≥99.0% (NT)

form

crystalline powder

mp

170-174 °C

solubility

H2O: 0.5 M at 20 °C, clear

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤10 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

SMILES string

OP(O)(O)=O.NC(CO)(CO)CO

InChI

1S/C4H11NO3.H3O4P/c5-4(1-6,2-7)3-8;1-5(2,3)4/h6-8H,1-3,5H2;(H3,1,2,3,4)

InChI key

JLEXUIVKURIPFI-UHFFFAOYSA-N

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library