Skip to Content
MilliporeSigma

91963

Cytotoxicity detection kit

100 tests

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.61
UNSPSC Code:
41116133
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

usage

100 tests

storage temp.

−20°C

General description

Cytotoxicity LDH Assay Kit-WST is a kit for determination of cytotoxicity by measuring a lactate dehydrogenase (LDH) activity released from damaged cells. LDH is a stable cytoplasmic enzyme present in all types of cells and released into the cell culture medium through damaged plasma membrane. Since Cytotoxicity LDH Assay kit-WST neither reflects the activity of living cells nor is harmful to cells, cytotoxicity can be measured with living cells (homogeneous assay). In addition, it is possible to perform a non-homogeneous assay by using the cell culture supernatant.

After preparation the Working Solution is stable under refrigerated conditions and can readily be used for up to six months.

Features and Benefits

  • Colorimetric assay system
  • Utilize for both homo- and non-homogeneous assays
  • Can be used with and without transferring supernatant
  • Stable Working Solution (6 months at 0°C)


pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
91963-1KT04061835535729