Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
MDL number:
form
solution
Quality Segment
IVD
for in vitro diagnostic use
concentration
15 mg/mL
pH
<1.0
application(s)
hematology
histology
shipped in
wet ice
storage temp.
2-8°C
SMILES string
Cc1cc(NC(=O)c2ccccc2)c(Cl)cc1N
InChI
1S/C14H13ClN2O/c1-9-7-13(11(15)8-12(9)16)17-14(18)10-5-3-2-4-6-10/h2-8H,16H2,1H3,(H,17,18)
InChI key
OIDXADKOPPNJOB-UHFFFAOYSA-N
Application
Used as a kit component in Procedure 91 for the demonstration of specific esterase activity in peripheral blood smears or paraffin sections.
Other Notes
Fast Red violet LB base, 15 mg/mL in 0.4 mol/L hydrochloric acid with stabilizer
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Doaa Ibrahim et al.
Animals : an open access journal from MDPI, 9(6) (2019-06-20)
This study compares between different selenium forms (sodium selenite; SeS, selenomethionine; Met-Se or nano-Se) and levels on growth performance, Se retention, antioxidative potential of fresh and frozen meat, and genes related to oxidative stress in Ross broilers. Birds (n =
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SML1086-25MG | 04061832451466 |
| SML1086-5MG | 04061832451473 |
| 36580-25G-F | 04061831825886 |
| 912-3ML | 04061837280108 |
| 912-10ML | 04061838203557 |
| 921718-1EA | 04065266541007 |
| 921971-1EA | 04065266694628 |
| 412198-250ML | 04061837406515 |
| 412198-1L | 04061837406508 |

