Skip to Content
MilliporeSigma

903C

TRIZMAL™ 6.3 Buffer Concentrate

pH 6.3(concentrate), 0.2 M

Synonym(s):

Trizmal buffer, Trizma® maleate buffer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352106
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Suitable for use in the absence of visible microbial growth.

contains

(Chloroform added as preservative)

IVD

for in vitro diagnostic use

concentration

0.2 M

pH

6.3(concentrate)

application(s)

hematology
histology

storage temp.

room temp

Application

For use in the Naphthol AS-D Chloroacetate Kit No. 90-C2

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany
Trizmal is a trademark of Sigma-Aldrich Co. LLC


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)

pictograms

Health hazardEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

target_organs

Central nervous system



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library