Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
biological source
plant (Clematis cultivar)
Quality Segment
assay
≥98.0% (HPLC)
form
powder or crystals
technique(s)
HPLC: suitable
color
light yellow
mp
220 - 225 °C ((428 - 437 °F ))
storage temp.
−20°C
SMILES string
C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(O)cc(O)c4C3=O)c5ccc(O)cc5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O
InChI
1S/C27H30O15/c1-9-17(31)20(34)22(36)26(39-9)38-8-15-18(32)21(35)23(37)27(41-15)42-25-19(33)16-13(30)6-12(29)7-14(16)40-24(25)10-2-4-11(28)5-3-10/h2-7,9,15,17-18,20-23,26-32,34-37H,8H2,1H3/t9-,15+,17-,18+,20+,21-,22+,23+,26+,27-/m0/s1
InChI key
RTATXGUCZHCSNG-QHWHWDPRSA-N
General description
Kaempferol 3-O-β -rutinoside is a bitter tasting flavonol glycoside that was isolated from flowers of a Clematis cultivar.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 90242-1MG | 04061833246511 |
| 90242-5MG | 04061833246528 |