Skip to Content
MilliporeSigma

90190

Transferrin from human blood plasma

≥95% (GE)

Synonym(s):

Human transferrin, Siderophilin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
234-318-8
NACRES:
NA.61
MDL number:
eCl@ss:
34058011
Form:
powder or crystals
Assay:
≥95% (GE)
Biological source:
human plasma
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human plasma

Quality Segment

assay

≥95% (GE)

form

powder or crystals

technique(s)

tissue culture: suitable

impurities

≤0.01% iron traces (AAS)

solubility

H2O: 50 mg/mL, clear, colorless to faintly yellow or pink

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... TF(7018)

Application

Transferrin is an important supplement in serum-free tissue culture media.

Biochem/physiol Actions

Transferrin is glycoprotein, approx. 80 kDa, with two binding sites for iron. It transports iron from its primary store in the liver to tissues that require iron, primarily bone marrow and spleen, but also brain. It binds Fe3+ extremely tightly (Kdiss 10-30 to 10-27), protecting against oxidant effects of free plasma iron.

Other Notes

Review: Iron transport and storage

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library