Skip to Content
MilliporeSigma

89862

β-1,3-Glucan from Euglena gracilis

Synonym(s):

Paramylon

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Euglena gracilis)

Quality Segment

form

powder

color

white to faint beige

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O[C@@H]3O[C@@H]([C@H](C[C@H]3O)O)CO)O)O[C@H]2[C@@H]([C@H](OC[C@@H]2O)CO)O

InChI

1S/C18H32O14/c19-2-9-6(22)1-7(23)17(29-9)32-16-13(26)11(4-21)30-18(14(16)27)31-15-8(24)5-28-10(3-20)12(15)25/h6-27H,1-5H2/t6-,7+,8-,9+,10+,11+,12+,13+,14+,15+,16-,17-,18-/m0/s1

InChI key

SPMCUTIDVYCGCK-IIIGWGBSSA-N

General description

The molecule consists of a linear chain of glucose units without any branching.

Application

β-1,3-Glucan from Euglena gracilis may be compared with other β-1,3-glucans and their derivatives to study mechanisms of protection of DNA from oxidative lesions. β-1,3-Glucan from Euglena gracilis may be used as a substrate to identify, differentiate and characterize glucanase(s).

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library