Skip to Content
MilliporeSigma
All Photos(1)

Documents

89478

Sigma-Aldrich

Fluorescent IEF-Marker pI 5.9 solution

2 mg/mL in 10 mM HCl

Synonym(s):

IEF-Marker fluorescent pI 5.9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105336
NACRES:
NA.26

mol wt

Mw 307.34 g/mol

Quality Level

concentration

2 mg/mL in 10 mM HCl

fluorescence

λex 330 nm; λem 415 nm (0.1 M citrate pH 5.9)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

2-8°C

General description

filtered through a 0.45 μm membrane filter

Application

suitable as IEF-Marker for HPCE and for IEF-gels

Reconstitution

For use in HPCE the marker should be diluted 1:100 in a suitable ampholyte solution (e.g. 2% ampholyte 3-10), UV-detection at 280 nm (20 °C). For use in IEF-gels the solution can be loaded directly onto the gel (max. 1 μl per lane).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Information on Isoelectric Focusing including what it is and how it is used. In order to ensure the high performance of analysis, isoelectric point (pI) standards are needed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service