Skip to Content
MilliporeSigma

88997

6-Carboxy-tetramethylrhodamine N-succinimidyl ester

BioReagent, suitable for fluorescence

Synonym(s):

"5-Carboxytetramethylrhodamine N-succinimidyl ester"

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H25N3O7
CAS Number:
Molecular Weight:
527.52
UNSPSC Code:
12352202
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

form

powder

solubility

DMF: 2 mg/mL, clear, acetonitrile: soluble

fluorescence

λex 543 nm; λem 575 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[N+](=[c]1cc2[o]c3c(c(c2cc1)c4c(ccc(c4)C(=O)ON5C(=O)CCC5=O)C(=O)[O-])ccc(c3)N(C)C)(C)C

InChI

1S/C29H25N3O7/c1-30(2)17-6-9-20-23(14-17)38-24-15-18(31(3)4)7-10-21(24)27(20)22-13-16(5-8-19(22)28(35)36)29(37)39-32-25(33)11-12-26(32)34/h5-10,13-15H,11-12H2,1-4H3

InChI key

PAOQTZWNYMMSEE-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Fluorescent labeling of peptides after chemical modification; Fluorescent labeling of aminoglycoside antibiotics


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library