Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88021102

Chang Liver (HeLa derivative) cell line

human cervix, Epithelial

Synonym(s):

Chang Liver cell line (HeLa derivative)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Chang Liver (HeLa derivative) cell line,

biological source

human cervix

form

liquid

growth mode

Adherent

karyotype

Modal no. 70

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix carcinoma

Cell Line Description

The cell line Chang Liver was originally thought to be derived from normal liver. The cell line has since been found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line should be considered as derived from HeLa. Ethnicity: Black.

Application

Chang Liver (HeLa derivative) cell line has been used to study the construction and biological activity of human torque teno virus (TTV) genotype 6.
Virus, biochemical and replication studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 12,14
D16S539: 9,10
D5S818: 12
D7S820: 8,12
THO1: 7
TPOX: 8,12
vWA: 16,18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clonal variation and interaction of cells with viruses.
W H MURPHY et al.
National Cancer Institute monograph, 7, 249-271 (1962-04-01)
Construction and biological activity of a full?length molecular clone of human Torque teno virus (TTV) genotype 6
Kakkola L, et al.
FEBS Journal, 274(18), 4719-4730 (2007)
Kofi D Kwofie et al.
Antimicrobial agents and chemotherapy, 60(6), 3283-3290 (2016-03-10)
Trypanosoma brucei parasites are kinetoplastid protozoa that devastate the health and economic well-being of millions of people in Africa through the disease human African trypanosomiasis (HAT). New chemotherapy has been eagerly awaited due to severe side effects and the drug
Continuous subcultivation of epithelial-like cells from normal human tissues.
R S M CHANG
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 87(2), 440-443 (1954-11-01)
Takuhiro Uto et al.
The American journal of Chinese medicine, 43(4), 757-767 (2015-06-30)
Alnus japonica Steud is a tree that grows in damp areas of mountain valleys and has been used as a traditional medicine in Asia. We investigated the antiproliferative activity of hirsutanone (Hir) and oregonin (Ore) in human cancer cell lines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service