Skip to Content
MilliporeSigma

Skip To

86454

TERGITOL TMN 10

Non-ionic, 1313 ppm, liquid, cloud point 76 °C

Synonym(s):

Trimethylnonylpolyethylene glycol, Polyethylene glycol trimethylnonyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

non-ionic

form

liquid

mol wt

~683 g/mol

impurities

~10% water

CMC

1313 ppm

HLB value

~14.1

transition temp

cloud point 76 °C

density

1.04 g/mL at 20 °C (lit.)

HLB

14.4

SMILES string

O(C(CC(CC(C)C)C)CC(C)C)CCO

InChI

1S/C14H30O2/c1-11(2)8-13(5)10-14(9-12(3)4)16-7-6-15/h11-15H,6-10H2,1-5H3

InChI key

VKKFWRYCMZBXIG-UHFFFAOYSA-N

General description

TERGITOL TMN 10, a heterogeneous non-ionic amphiphilic surfactant has a hydrophobic moiety of 2,6,8-trimethyl-4-nonanol and a hydrophilic side chain of polyoxyethylene glycol with an average of 10 ethylene oxide units. This alkyl-substituted surfactant is soluble in water and most polar organic solvents.[1]

Application


  • Decomposition of non-ionic surfactant Tergitol TMN-10 by the Fenton process in the presence of iron oxide nanoparticles.: This study explores the degradation of Tergitol TMN-10 using the Fenton process, enhanced by iron oxide nanoparticles, highlighting its potential for environmental remediation strategies (Kos et al., 2014).

  • Application of nonionic polyoxyethylene detergents in studies of phosphatidylinositol kinase.: This research investigates the utility of nonionic polyoxyethylene detergents, such as Tergitol TMN-10, in enzymatic studies, specifically focusing on phosphatidylinositol kinase activity, which is crucial for understanding cellular signaling mechanisms (Ganong and Lu, 1989).


Biochem/physiol Actions

Tergitol series surfactants are polyethoxylated secondary alcohols[1] that can be used as foam-controlling agents.

Features and Benefits

  • Suitable for use as a high-temperature penetrant & dispersant
  • High hydrophilic-lipophilic balance (HLB) emulsifier
  • Features narrow gel range
  • Biodegradable[1]

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
86453NP10NP40S
form

liquid

form

liquid

form

-

form

liquid

description

non-ionic

description

non-ionic

description

non-ionic

description

non-ionic

impurities

~10% water

impurities

~10% water

impurities

-

impurities

-

density

1.04 g/mL at 20 °C (lit.)

density

1.01 g/mL at 20 °C

density

-

density

(1,105 g/cm3 at 20 °C)

HLB

14.4

HLB

13.1

HLB

13.2

HLB

17.8

CMC

1313 ppm

CMC

800 ppm

CMC

55 ppm

CMC

232 mg/L (25°C)


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

266.0 °F - closed cup

flash_point_c

130 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters