Skip to Content
MilliporeSigma

Skip To

81189

Poly(ethylene glycol)

greener alternative

Greener Alternative

BioUltra, Molecular Biology, 1,000

Synonym(s):

Polyethylene glycol, PEG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352104
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

product line

BioUltra

form

solid

mol wt

Mr 950-1050

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

solubility

H2O: 50 mg/mL at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: ≤0.10, λ: 280 nm Amax: ≤0.05

greener alternative category

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. Polyethylene glycol (PEG) is an eco-friendly, biodegradable polymer widely used in pharmaceuticals and cosmetics. Its non-toxic nature and versatility make it a sustainable choice, derived from renewable resources, contributing to greener product formulations. Click here for more information.
Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system, which is required for the purification of biological materials. PEG also interacts with cell membrane, thereby allowing cell fusion.

Application

Poly(ethylene glycol) has been used to develop a method for efficient transformation of Brevibacterium lactofermentum protoplasts with plasmid DNA. It has been used as a fusion agent to fuse spleen cells with the myeloma cell line SP2/0.

Other Notes

PEG precipitation of DNA fragments

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
812688844084797
description

BioUltra, Molecular Biology, 1,000

description

BioUltra, Molecular Biology, 8,000

description

BioUltra, 200

description

BioUltra, 2,000

form

solid

form

powder

form

liquid

form

powder

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

mol wt

Mr 950-1050

mol wt

Mr 7000-9000

mol wt

Mr 190-210

mol wt

Mr 1900-2200

cation traces

Al: ≤5 mg/kg, Ba: ≤5 mg/kg, Ca: ≤10 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, K: ≤200 mg/kg, Mg: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Sr: ≤5 mg/kg, As: ≤0.1 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Mn: ≤5 mg/kg, Pb: ≤5 mg/kg, Bi: ≤5 mg/kg, Li: ≤5 mg/kg, Zn: ≤5 mg/kg, Cr: ≤5 mg/kg, Na: ≤200 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤500 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

impurities

DNases, none detected, insoluble matter, passes filter test, proteases, none detected, RNases, none detected, phosphatases, none detected

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected

impurities

insoluble matter:, passes filter test, ≤0.02% peroxides (as H2O2):

impurities

insoluble matter:, passes filter test, ≤0.001% peroxides (as H2O2):

UV absorption

λ: 260 nm Amax: ≤0.10, λ: 280 nm Amax: ≤0.05

UV absorption

λ: 260 nm Amax: 0.15, λ: 280 nm Amax: 0.10

UV absorption

λ: 260 nm Amax: 0.06, λ: 280 nm Amax: 0.03

UV absorption

λ: 260 nm Amax: ≤0.1, λ: 280 nm Amax: ≤0.04


Still not finding the right product?

Explore all of our products under Poly(ethylene glycol)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters