Skip to Content

76755

Pentafluorophenyl isothiocyanate

≥97.0% (GC)

Synonym(s):

1,2,3,4,5-pentafluoro-6-isothiocyanatobenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7F5NS
CAS Number:
Molecular Weight:
225.14
UNSPSC Code:
12352200
NACRES:
NA.25
PubChem Substance ID:
EC Number:
252-791-9
Beilstein/REAXYS Number:
2736207
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.540

bp

38-39 °C/0.2 mmHg (lit.)

density

1.594 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(N=C=S)c(F)c1F

InChI

1S/C7F5NS/c8-2-3(9)5(11)7(13-1-14)6(12)4(2)10

InChI key

NGNKMRBGZPDABE-UHFFFAOYSA-N



pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library