Skip to Content
MilliporeSigma

74931

Alcohol Dehydrogenase

≥10.0 U/mg

Synonym(s):

ADH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.77
EC Number:
MDL number:
Specific activity:
≥10.0 U/mg
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

specific activity

≥10.0 U/mg

storage temp.

−20°C

Other Notes

1 U corresponds to the amount of enzyme which reduces 1 μmol 3-Nonanone per minute at pH 7.0 and 37°C


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library