Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74675

Sigma-Aldrich

Novobiocin sodium salt

≥93% (HPLC)

Synonym(s):

Albamycin, Cathomycin, Novobiocin, Novobiocin, Monosodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H35N2NaO11
CAS Number:
Molecular Weight:
634.61
Beilstein:
3892910
EC Number:
UNSPSC Code:
51286504
NACRES:
NA.85

Assay

≥93% (HPLC)

form

powder

optical activity

[α]20/D −48±3°, c = 1% in ethanol

color

white to faint beige

solubility

H2O: soluble

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

[Na+].CO[C@@H]1[C@@H](OC(N)=O)[C@@H](O)[C@H](Oc2ccc3C([O-])=C(NC(=O)c4ccc(O)c(C\C=C(\C)C)c4)C(=O)Oc3c2C)OC1(C)C

InChI

1S/C31H35N2O11.Na/c1-14(2)7-8-16-13-17(9-11-19(16)34)27(37)33-21-22(35)18-10-12-20(15(3)24(18)42-28(21)38)41-29-23(36)25(43-30(32)39)26(40-6)31(4,5)44-29;/h7,9-13,23,25-26,29,34,36H,8H2,1-6H3,(H2,32,39)(H,33,37);/q-1;+1

InChI key

AXOUUAINTJNFRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: coumarin-glycoside

Application

For the production of positively supercoiled plasmid DNA. Inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase. Inhibitor of retrovirus RNA-dependent DNA-polymerase.
Novobiocin is used to study heat shock protein inhibition and to produce positively supercoiled plasmid DNA. It is an inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase. It also is an inhibitor of retrovirus RNA-dependent DNA-polymerase.

Biochem/physiol Actions

Mode of Action: Inhibits DNA synthesis by inhibiting the enzyme Topoisomerase II.
Antimicrobial spectrum: Gram-positive bacterial.
Novobiocin inhibits DNA synthesis by inhibiting bacterial DNA gyrase targeting the GyrB subunit of the enzyme involved in energy transduction. Novobiocin acts as a competitive inhibitors of the ATPase reaction catalysed by GyrB . It is also a Hsp90 inhibitor . Antimicrobial spectrum: Gram-positive bacterial.

Packaging

1g, 5g

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Store under inert gas. Keep in a dry place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Sumiyoshi et al.
The Journal of general virology, 64 (Pt 10), 2329-2333 (1983-10-01)
Inhibitors of bacterial DNA gyrase and eukaryotic DNA topoisomerase (novobiocin and nalidixic acid) were investigated with respect to their effect on the activity of RNA-dependent DNA polymerases from murine and avian retroviruses. Purified RNA-dependent DNA polymerase from AKR virus was
N R Cozzarelli
Science (New York, N.Y.), 207(4434), 953-960 (1980-02-29)
Negative supercoiling of bacterial DNA by DNA gyrase influences all metabolic processes involving DNA and is essential for replication. Gyrase supercoils DNA by a mechanism called sign inversion, whereby a positive supercoil is directly inverted to a negative one by
D Lockshon et al.
Nucleic acids research, 11(10), 2999-3017 (1983-05-25)
A procedure has been developed whereby the relative amounts of the topoisomers of E. coli plasmid can be determined for cells grown under a variety of conditions. Several applications of the procedure are presented. Addition of either novobiocin or oxolinic
A Maxwell
Molecular microbiology, 9(4), 681-686 (1993-08-01)
The coumarin group of antibiotics have as their target the bacterial enzyme DNA gyrase. The drugs bind to the B subunit of gyrase and inhibit DNA supercoiling by blocking the ATPase activity. Recent data show that the binding site for
Lorenzo González-Molleda et al.
Antimicrobial agents and chemotherapy, 56(2), 893-902 (2011-11-23)
The lytic DNA replication of Kaposi's sarcoma-associated herpesvirus (KSHV) initiates at an origin (ori-Lyt) and requires trans-acting elements, both viral and cellular. We recently demonstrated that several host cellular proteins, including topoisomerases I and II (Topo I and II), are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service