Skip to Content

73873

Lectin from Ulex europaeus-Atto 594 conjugate

Synonym(s):

Ulex europaeus agglutinin-Atto 594 conjugate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352108
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

carbohydrate specificity α-L-Fucose

Quality Segment

form

powder

fluorescence

λex 601 nm; λem 627 nm in PBS

storage temp.

−20°C

General description

Lectins are ubiquitous proteins or glycoproteins with a high and specific affinity to sugar residues. They allow a sensitive detection of cellular carbohydrates, revealing subtle alteration in glycosylation between otherwise indistinguishable cells.
Atto-dyes have very bright fluorescent signals and high photostability, which enable a direct one-step tissue-binding protocol. Time-consuming multistage amplification procedures are not required for Atto-dye lectin conjugates

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library