Quality Segment
form
aqueous suspension
concentration
2.5% (solids)
surface coverage
reactive groups (COOH) >0.1 mmol/g polymer
particle size
1 μm std dev <0.1 μm, coeff var <5%
Analysis Note
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer. They are printed on the individual certificates.
Extinction maximum: 633 nm
Emission maximum: 672 nm
Extinction maximum: 633 nm
Emission maximum: 672 nm
Still not finding the right product?
Explore all of our products under Micro particles based on melamine resin, carboxylate-modified, nile blue-marked
Storage Class
10 - Combustible liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.