Skip to Content
MilliporeSigma

69579

γ,γ-Dimethylallyl pyrophosphate ammonium salt

≥93.0% (TLC)

Synonym(s):

3-Methyl-2-butenyl pyrophosphate ammonium salt, DMAPP, Prenyl pyrophosphate ammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H12O7P2 · xNH3
CAS Number:
Molecular Weight:
246.09 (free acid basis)
UNSPSC Code:
12352205
NACRES:
NA.25
Assay:
≥93.0% (TLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥93.0% (TLC)

form

powder

storage temp.

−20°C

SMILES string

N.C\C(C)=C\COP(O)(=O)OP(O)(O)=O

InChI

1S/C5H12O7P2.H3N/c1-5(2)3-4-11-14(9,10)12-13(6,7)8;/h3H,4H2,1-2H3,(H,9,10)(H2,6,7,8);1H3

InChI key

DPJANMSJRVSJKR-UHFFFAOYSA-N

Application

Prenyl pyrophosphate, an intermediate in mevalonic acid pathway isoprenoid/terpene biosynthesis, is use to study the kinetics of farnesyl pyrophosphate synthases.

Biochem/physiol Actions

Intermediate in terpene biosynthesis

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library