Skip to Content

68606

Atto 594-Streptavidin

BioReagent, suitable for fluorescence

Synonym(s):

Streptavidin-Atto 594

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352108
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

form

solid

manufacturer/tradename

ATTO-TEC GmbH

λ

in PBS (buffer)

UV absorption

λ: 604-610 nm Amax

suitability

corresponds for SDS-PAGE, suitable for fluorescence

storage temp.

−20°C

General description

Atto 594 is a novel fluorescent label belonging to the class of Rhodamine dyes. The dye is designed for application in the area of life science, e.g. labeling of DNA, RNA or proteins. Characteristic features of the label are strong absorption, high fluorescence quantum yield, high thermal and photo-stability, excellent water solubility, and very little triplet formation. After coupling to a substrate Atto 594 carries a net electrical charge of -1.

find more information here

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

dye-to-protein ration (molar) ≥1.5

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library