Skip to Content
MilliporeSigma

64605

Methyl cellulose

27.5-31.5% methoxyl basis

Synonym(s):

Methyl cellulose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

concentration

27.5-31.5% (methoxyl)

color

white to off-white

viscosity

10-25 mPa.s, 2 % in H2O(20 °C)

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Methocel is a registered trademark of The Dow Chemical Company or an affiliated company of Dow


Still not finding the right product?

Explore all of our products under Methyl cellulose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
M7753-50MG04061834062363
M7753-500MG04061834062356
M7753-100MG04061834062349
64605-500G-F04061835365807
64605-100G-F04061835365791