Skip to Content
MilliporeSigma

63138

Magnesium sulfate heptahydrate

BioUltra, ≥99.5% (KT)

Synonym(s):

Epsom salts

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
PubChem Substance ID:
eCl@ss:
38030409
UNSPSC Code:
12352302
NACRES:
NA.26
EC Number:
231-298-2
MDL number:
Assay:
≥99.5% (KT)
Grade:
reagent grade
Solubility:
H2O: 1 M at 20 °C, clear, colorless
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reagent grade

Quality Segment

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

BioUltra

assay

≥99.5% (KT)

impurities

insoluble matter, passes filter test, ≤0.002% total nitrogen (N)

pH

5.0-8.0 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤5 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤1 mg/kg, K: ≤10 mg/kg, Li: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤10 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.010, λ: 280 nm Amax: 0.010

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Application


  • Research on the Working Performance and the Corresponding Mechanical Strength of Polyaluminum Sulfate Early Strength Alkali-Free Liquid Accelerator Matrix Cement.: This research investigates the applications of magnesium sulfate heptahydrate in improving the performance and mechanical strength of cement used in construction (Wang et al., 2022).

Other Notes

Component of Earle′s balanced salt solution; Component of Hanks′ balanced salt solution; Component of plant cell-culture media and of culture media for microorganisms.; Purification of CAP, catabolite gene activator; In the preparation of spheroplasts from E. coli K38


Still not finding the right product?

Explore all of our products under Magnesium sulfate heptahydrate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library