Skip to Content
MilliporeSigma

62612

Lithium sulfate monohydrate

BioUltra, ≥99.0% (T)

Synonym(s):

Lithium sulfate (Li2SO4) monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Li2SO4 · H2O
CAS Number:
Molecular Weight:
127.96
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
233-820-4
MDL number:
Assay:
≥99.0% (T)
Grade:
reagent grade
Form:
solid
Solubility:
H2O: 1 M at 20 °C, clear, colorless
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reagent grade

Quality Segment

product line

BioUltra

assay

≥99.0% (T)

form

solid

impurities

insoluble matter, passes filter test

pH

4-7 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, nitrate (NO3-): ≤10 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.3, λ: 280 nm Amax: ≤0.1

SMILES string

[Li+].[Li+].O.[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2

InChI key

RKGLUDFWIKNKMX-UHFFFAOYSA-L

Application


  • Crystallographic studies in molecular biology: Lithium sulfate monohydrate is crucial in the preparation of crystal structures for biochemical assays, facilitating the deciphering of complex biological structures such as the novel NEIL1 nanobody, which has significant implications for genetic research and pharmaceutical applications (Thompson et al., 2023).

  • Enhanced nuclear magnetic resonance spectroscopy: Utilized in multiple-quantum magic-angle spinning spectroscopy, lithium sulfate monohydrate improves the resolution and quality of NMR studies. This application is vital for advancing molecular characterization techniques used in pharmaceutical synthesis and material science research (Rovnyak et al., 2003).



Still not finding the right product?

Explore all of our products under Lithium sulfate monohydrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library