Skip to Content
MilliporeSigma

61367

Myristoyl-L-carnitine

≥97.0% (HPLC)

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxotetradecyl)oxy]-1-propanaminium inner salt, L-Carnitine tetradecanoyl ester, C14-Carnitine, Tetradecanoyl-L-carnitine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H41NO4
CAS Number:
Molecular Weight:
371.55
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.26
Beilstein/REAXYS Number:
4149853
Assay:
≥97.0% (HPLC)
Storage condition:
under inert gas (Argon)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (HPLC)

optical activity

[α]/D -18±2°, c = 1 in methanol

storage condition

under inert gas (Argon)

impurities

≤10% water

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCCCCCCCC)=O)CC([O-])=O

InChI

1S/C21H41NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4/h19H,5-18H2,1-4H3/t19-/m1/s1

InChI key

PSHXNVGSVNEJBD-LJQANCHMSA-N

Biochem/physiol Actions

Myristoyl-L-carnitine is a human carnitine involved in beta-oxidation of long-chain fatty acids.


Still not finding the right product?

Explore all of our products under Myristoyl-L-carnitine


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library