Skip to Content
MilliporeSigma
All Photos(1)

Documents

59300C

SAFC

Dulbecco′s Phosphate Buffered Saline

liquid, sterile-filtered, with calcium, with magnesium, suitable for cell culture

Pharma Manufacturing

Synonym(s):

DPBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Dulbecco′s Phosphate Buffered Saline (DPBS) is intended for use in the maintenance of mammalian cells where a chemically defined balanced salt solution provides an environment that will maintain the structural and physiological integrity of cells in vitro.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louis-Marie Bloyet et al.
Journal of virology, 94(13) (2020-04-24)
Nonsegmented negative-strand (NNS) RNA viruses possess a ribonucleoprotein template in which the genomic RNA is sequestered within a homopolymer of nucleocapsid protein (N). The viral RNA-dependent RNA polymerase (RdRP) resides within an approximately 250-kDa large protein (L), along with unconventional
Simon Jenni et al.
Cell reports, 30(1), 53-60 (2020-01-09)
The large (L) proteins of non-segmented, negative-strand RNA viruses are multifunctional enzymes that produce capped, methylated, and polyadenylated mRNA and replicate the viral genome. A phosphoprotein (P), required for efficient RNA-dependent RNA polymerization from the viral ribonucleoprotein (RNP) template, regulates
Teresa Herrera et al.
Food chemistry, 270, 509-517 (2018-09-04)
Ethanol extracts (EE) from fenugreek and quinoa seeds with different total content of inhibitory compounds (TIC, total saponin plus phenolic) were prepared with and without concentration of TIC (CEE -concentrated EE-, and EE, respectively). Their inhibitory activity on pancreatic lipase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service