Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59202C

SAFC

L-Glutamine Solution 200 mM

29.23 mg/mL in saline, solution, suitable for cell culture

Pharma Manufacturing

Synonym(s):

L-Glutamine solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

Quality Level

description

for research or for further manufacturing use

sterility

sterile; sterile-filtered

form

solution

technique(s)

cell culture | mammalian: suitable

components

L-glutamine: 29.23 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1

InChI key

ZDXPYRJPNDTMRX-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Leonetti et al.
Scientific reports, 9(1), 4591-4591 (2019-03-16)
Standard imaging systems provide a spatial resolution that is ultimately dictated by the numerical aperture (NA) of the illumination and collection optics. In biological tissues, the resolution is strongly affected by scattering, which limits the penetration depth to a few
Xiaolu Zhang et al.
Nature communications, 10(1), 4732-4732 (2019-10-20)
Current multiplexing strategies for massively parallel sequencing of genomic DNA mainly rely on library indexing in the final steps of library preparation. This procedure is costly and time-consuming, because a library must be generated separately for each sample. Furthermore, library
Jil Sander et al.
Immunity, 47(6), 1051-1066 (2017-12-21)
Human in vitro generated monocyte-derived dendritic cells (moDCs) and macrophages are used clinically, e.g., to induce immunity against cancer. However, their physiological counterparts, ontogeny, transcriptional regulation, and heterogeneity remains largely unknown, hampering their clinical use. High-dimensional techniques were used to elucidate transcriptional, phenotypic
Olga Z Karicheva et al.
Nucleic acids research, 39(18), 8173-8186 (2011-07-05)
Mutations in human mitochondrial DNA are often associated with incurable human neuromuscular diseases. Among these mutations, an important number have been identified in tRNA genes, including 29 in the gene MT-TL1 coding for the tRNA(Leu(UUR)). The m.3243A>G mutation was described
Muna Ibrahim et al.
Diabetologia, 63(1), 162-178 (2019-11-30)
Exposure to environmental pollution has been consistently linked to diabetes incidence in humans, but the potential causative mechanisms remain unclear. Given the critical role of regulated insulin secretion in maintaining glucose homeostasis, environmental chemicals that reach the endocrine pancreas and

Articles

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service