Skip to Content
MilliporeSigma
All Photos(1)

Documents

56395

Sigma-Aldrich

N-Hexanoyl-L-homoserine lactone

≥96% (HPLC)

Synonym(s):

N-Caproyl-L-homoserine lactone, N-[(3S)-Tetrahydro-2-oxo-3-furanyl]hexanamide, C6-HSL, HHL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
Beilstein:
8143287
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.28

product name

N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)

Quality Level

Assay

≥96% (HPLC)

form

powder or crystals

optical activity

[α]/D -32±3°, c = 0.2 in methanol

color

white to off-white

suitability

conforms to structure for Proton NMR spectrum

storage temp.

−20°C

SMILES string

O=C1OCC[C@@H]1NC(CCCCC)=O

InChI

1S/C10H17NO3/c1-2-3-4-5-9(12)11-8-6-7-14-10(8)13/h8H,2-7H2,1H3,(H,11,12)/t8-/m0/s1

InChI key

ZJFKKPDLNLCPNP-QMMMGPOBSA-N

Biochem/physiol Actions

N-Hexanoyl-L-homoserine lactone is a member of N-acyl-homoserine lactone family. N-Acylhomoserine lactones (AHL) regulate gene expression in gram-negative bacteria, such as Echerichia and Salmonella, and are involved in quorum sensing, cell to cell communication among bacteria; for reviews see. Bacterial intercellular communication has become a target for the development of new anti-virulence drugs, and a research focus for the prevention of biofilm formation.
Quorum-sensing signal generation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Torabi Delshad et al.
Journal of applied microbiology, 125(2), 356-369 (2018-04-26)
A variety of pathogens use quorum sensing (QS) to control the expression of their virulence factors. QS interference has hence been proposed as a promising antivirulence strategy. The specific aim of this study was to isolate bacteria from trout tissue
Jeng Yeong Chow et al.
Biochemistry, 48(20), 4344-4353 (2009-04-21)
The PLL(PTE-like lactonase)-group of enzymes within the amidohydrolase superfamily hydrolyze N-acyl-homoserine lactones (AHLs) that are involved in bacterial quorum-sensing pathways. These enzymes possess the (beta/alpha)(8)-barrel fold and serve as attractive templates for in vitro evolution and engineering of quorum-quenching biological
Chi-I Chang et al.
Molecules (Basel, Switzerland), 24(11) (2019-06-13)
Three new dimeric abietane-type diterpenoids, abieta-6,8,11,13-tetraen-12-yl 12-hydroxyabieta-8,11,13-trien-7α-yl peroxide (1), abieta-6,8,11,13-tetraen-12-yl 12-hydroxyabieta-8,11,13-trien-7β-yl peroxide (2), and 12-hydroxyabieta-8,11,13-trien-7β-yl 7-oxoabieta-5,8,11,13-tetraen-12-yl peroxide (3), together with four known abietane-type diterpenoids (4-7) were isolated from the methanol extract of the bark of Cryptomeria japonica. Their structures were
Xue Yang et al.
Ultrasonics sonochemistry, 38, 19-28 (2017-06-22)
The effects of ultrasound pretreatment with different frequencies and working modes, including mono-frequency ultrasound (MFU), dual-frequency ultrasound (DFU) and tri-frequency ultrasound (TFU), on the degree of hydrolysis (DH) of rice protein (RP) and angiotensin-I-converting enzyme (ACE) inhibitory activity of RP
Orrapun Selamassakul et al.
Journal of food science, 85(3), 707-717 (2020-02-12)
This study evaluated the biological properties of peptides from brown rice protein hydrolyzed by bromelain (eb-RPH) in relation to flavor characteristic. The fractionation into peptides < 1 kDa was observed to improve the DPPH, ABTS, and hydroxyl radical-scavenging activities (0.19, 2.28, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service