Skip to Content
MilliporeSigma

54290

Hydroxyethyl-cellulose

viscosity 90-160 cP, 5 % in H2O(25 °C)

Synonym(s):

Cellosize® EP-09

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

color

beige

viscosity

90-160 cP, 5 % in H2O(25 °C)

storage temp.

room temp

SMILES string

O1C(C(C(C(C1CO)OC)O)OCCO)OC2C(OC(C(C2O)O)OC4C(OC(C(C4O)O)C)CO)COC3OC(C(C(C3O)O)OC)CO

InChI

1S/C29H52O21/c1-10-15(34)16(35)24(13(8-33)45-10)49-28-20(39)18(37)25(50-29-26(43-5-4-30)21(40)23(42-3)12(7-32)47-29)14(48-28)9-44-27-19(38)17(36)22(41-2)11(6-31)46-27/h10-40H,4-9H2,1-3H3

InChI key

CWSZBVAUYPTXTG-UHFFFAOYSA-N

General description

Hydroxyethyl cellulose is a gelling and thickening agent derived from cellulose and is widely used in cosmetics, cleaning solutions and other household products. It is used with hydrophobic drugs in capsule formulations, to improve the drugs dissolution in the gastrointestinal fluids. This process is known as hydrophilization.

Application

Hydroxyethylcellulose is used as a gelling and thickening agent in the development of biostructures for the delivery of hydrophobic drugs. Hydroxyethylcellulose is used in the development of polymer networks and block copolymers useful in separation technology such as capillary electrophoresis and in biofilms and coatings. It is also used for encapsulation of cells in chitosan/glycerophosphate/hydroxyethyl cellulose (chitosan-GP/HEC) gels.

Other Notes

Network-forming polymer in capillary electrophoresis of DNA restriction fragments
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

CELLOSIZE is a registered trademark of The Dow Chemical Company or an affiliated company of Dow


Still not finding the right product?

Explore all of our products under Hydroxyethyl-cellulose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library